Learn More
Chemglass Life Sciences Blank Foam Inserts, Set of 2 (Recessed Platform Required) (Fig. 9)
SDP
Catalog No. 5000083210
Encompass
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.
Catalog No. 50-000-83210 Supplier Chemglass Life Sciences Supplier No. CLS4891009
NOTE: VORTEX-GENIE 1 MAY ONLY BE USED WITH POP-OFF CUP AND 3" PLATFORM.