Learn More
Grainger Blank Labels Blank 5 in W PK10
Catalog No. 502749901
Encompass_Preferred
The Brady Blank Labels, 5 in Character, are white, blank labels made of vinyl material and measure 5 inches tall. They are designed for use with the Brady label maker to create custom labels. These vinyl labels are smudge, water, chemical, and heat resistant. Each roll contains 250 labels and measures 2.25 inches wide with a 1 inch core. They feature permanent adhesive backing so labels stay adhered once applied. Recommended uses include wire and cable marking, equipment ID, and general purpose identification and labeling applications.
Catalog No. 50-274-9901 Supplier Grainger Supplier No. 14R088
Add to Cart
Add to Cart
The Brady Blank Labels, 5 in Character, are white, blank labels made of vinyl material and measure 5 inches tall. They are designed for use with the Brady label maker to create custom labels. These vinyl labels are smudge, water, chemical, and heat resistant. Each roll contains 250 labels and measures 2.25 inches wide with a 1 inch core. They feature permanent adhesive backing so labels stay adhered once applied. Recommended uses include wire and cable marking, equipment ID, and general purpose identification and labeling applications.