Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldon™ Blank Sterile Disks
SCI_ED

Catalog No. S28515 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S28515 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28515 Supplier Aldon™ Supplier No. XD1368
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to test effectiveness of particular antimicrobial agents.

  • Each cartridge contains 50 disks with reclosable cap
  • Disks are 6.35mm (0.25 in.) in diameter
  • DOT Info: Non-regulated

Specifications

Grade 7 to 14
Format Disk
Product Type Blank Sterile Disks
Diameter (English) Ring 0.25 in.
Quantity 1 Cartridge with 50 Discs
Color White
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.