Learn More
Accuform™ Blank Yellow Barricade Tape
Catalog No. 19119208 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
19-119-208 Yellow
1 options
Catalog No. 19-119-208 Supplier Accuform™ Supplier No. MPT200BLNKY
Add to Cart
Add to Cart
Accuform™ barricade tape temporarily sections-off hazards and danger zones quickly and effectively
  • Bold warning message continuously repeated length of roll
  • Lightweight polyethylene plastic is durable for indoor and outdoor environments
  • Easily cut and secure with adhesive tape, nail, staples, or tie-off
Specifications
Color Yellow
Thickness 3 mil
Height (English) 3 in.
Material Polyethylene
Length (English) 1000 ft.
Adhesive Type Non-adhesive
Type Barricade Tape