Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Blank Yellow Marking Tape

Catalog No. 19119212 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-119-212 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-119-212 Supplier Accuform™ Supplier No. PTM623YL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ marking tape used to mark specific areas and promote safety by identifying potential hazard areas

  • Vinyl adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery, and equipment
  • Waterproof, wear-resistant, minor resistance to acids, oils, grease, caustics, alkalis, and most solvents
  • Permanent self-adhesive back with no liner

Specifications

Adhesive Type Permanent Adhesive
Displayed Text None
Height (English) 2 in.
Shape Rectangular
Color Yellow
Product Type Hazard Marking Tape
Length (English) 108 ft.
Material Vinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.