Learn More
Hach Company Blanking Reagent for ULR Chlorine, 29 mL
Supplier:  Hach Company 2493023
Encompass
For determination of ultra-low range chlorine by the DPD ULR Method. Range 2 - 500 µg/L, USEPA accepted for reporting wastewater analysis results.*This product has not been evaluated to test for chlorine and chlormaines in medical applications in the United States.
Catalog No. 50-199-6071