Learn More
Cayman Chemical BlarcamesIn 50mg
SDP
Catalog No. 502909743
Encompass_Preferred
A σ1 receptor agonist; inhibits reductions in mitochondrial oxygen consumption and lipid peroxidation induced by Aβ (25-35) in isolated mouse hippocampus at 1 mg/kg; improves deficits in novel object recognition induced by Aβ (25-35) in mice, as well as reduces tau phosphorylation and Aβ (1-42) levels in the hippocampus in the same model at 1 mg/kg; increases the latency to fall in the rotarod test, reduces gait impairments, and reduces increases in the mean acoustic startle response in an Mecp2-/- mouse model of Rett syndrome at 30 mg/kg per day
Catalog No. 50-290-9743 Supplier Cayman Chemical Supplier No. 3681250 MG
A σ1 receptor agonist; inhibits reductions in mitochondrial oxygen consumption and lipid peroxidation induced by Aβ (25-35) in isolated mouse hippocampus at 1 mg/kg; improves deficits in novel object recognition induced by Aβ (25-35) in mice, as well as reduces tau phosphorylation and Aβ (1-42) levels in the hippocampus in the same model at 1 mg/kg; increases the latency to fall in the rotarod test, reduces gait impairments, and reduces increases in the mean acoustic startle response in an Mecp2-/- mouse model of Rett syndrome at 30 mg/kg per day