Learn More
Medchemexpress LLC Blarcamesine hydrochloride | 195615-84-0 | 99.87% | 317.85 | 10 MG
SDP
Catalog No. 5000364691
Encompass_Preferred
Blarcamesine hydrochloride, also known as Anavex 2-73, is a Sigma-1 Receptor agonist. This compound is widely utilized in laboratory research, particularly for studies related to neurological and neurodegenerative diseases. It has also been explored in clinical trials for conditions such as breast cancer.
  • Acts as a Sigma-1 Receptor agonist with an IC50 of 860 nM
  • Exhibits a purity of 99.87%
  • Presents as a white to off-white solid
  • Offers solubility in H2O and DMSO
  • Suitable for various in vitro and in vivo research applications
Catalog No. 50-003-64691 Supplier Medchemexpress LLC Supplier No. 10MGHY101864
Blarcamesine hydrochloride, also known as Anavex 2-73, is a Sigma-1 Receptor agonist. This compound is widely utilized in laboratory research, particularly for studies related to neurological and neurodegenerative diseases. It has also been explored in clinical trials for conditions such as breast cancer.
  • Acts as a Sigma-1 Receptor agonist with an IC50 of 860 nM
  • Exhibits a purity of 99.87%
  • Presents as a white to off-white solid
  • Offers solubility in H2O and DMSO
  • Suitable for various in vitro and in vivo research applications