Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Blarcamesine hydrochloride | 195615-84-0 | MFCD30146449 | 99.9% | 317.85 g/mol | C19H24ClNO | 100 MG
SDP

Supplier:  Medchemexpress LLC HY101864100MG

Encompass_Preferred

Blarcamesine hydrochloride is the hydrochloride salt of blarcamesine, an investigational small-molecule sigma-1 receptor agonist supplied for research and preclinical studies. It is provided as a high-purity solid with reported in vitro activity and specific storage recommendations. Not for human or clinical use.

  • High purity solid suitable for in vitro and preclinical research.
  • Sigma-1 receptor agonist with reported IC50 of 860 nM.
  • Molecular formula C19H24ClNO and molecular weight 317.85 g/mol.
  • White to off-white appearance for easy identification.
  • Storage recommendations: store sealed, away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human use.

Catalog No. 50-002-48027


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.