Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Blast Off! Deluxe Kit Components
SCI_ED

Catalog No. S396045BO Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Balloons
Crucible tongs
Petri dish
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S396045BO Petri dish
S396085BO Crucible tongs
S051515BO Balloons
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S396045BO
Add to Cart
Add to Cart

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Grade K to 5
Dimensions (L x W) 100 x 25 mm
For Use With (Application) Education Kit
Material Polystyrene
Product Type Petri dish
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.