Learn More
Cayman Chemical Blasticidin A, Purity: ≥95%, CAS: 100513-53-9, 5 mg
SDP
Catalog No. 503015762
Encompass_Preferred
A bacterial metabolite; inhibits aflatoxin production by A. parasiticus without affecting fungal growth (IC50s = 0.25 and 1.6 μM, respectively); decreases ribosomal protein levels and inhibits protein synthesis in S. cerevisiae
Catalog No. 50-301-5762 Supplier Cayman Chemical Supplier No. 255195 MG
A bacterial metabolite; inhibits aflatoxin production by A. parasiticus without affecting fungal growth (IC50s = 0.25 and 1.6 μM, respectively); decreases ribosomal protein levels and inhibits protein synthesis in S. cerevisiae