Learn More
Invivogen Blasticidin | 2079-00-7 | MFCD02091639 | 500 mg, 50 mL/bottle
SDP
Catalog No. NC9411535
Encompass
Blasticidin | Purity: 95% | Mol Wt: 422.45 | 2079-00-7 | MFCD02091639 | 500 mg, 50 mL/bottle
Catalog No. NC9411535 Supplier Invivogen Supplier No. ANTBL5B
Blasticidin | Purity: 95% | Mol Wt: 422.45 | 2079-00-7 | MFCD02091639 | 500 mg, 50 mL/bottle