Learn More
AG Scientific Inc Blasticidin S HCl, 100 MG
Supplier:  AG Scientific Inc B1247100MG
Encompass_Preferred
Blasticidin S HCL is an antibiotic that demonstrates a strong inhibitory effect on protein synthesis in eukaryotic and prokaryotic cells. Blasticidin causes tRNA to bond more strongly to the P site of the large ribosomal subunit while simultaneously deforming the tRNA, thus inhibiting proper translation. The ultimate result of this process is a significant suppression of the hydrolysis of peptidyl-tRNA along with inhibition of peptide bond formation. Isolated from Streptomyces griseochromogenes. CAS Number: 3513-03-9Molecular Weight: 458.904Chemical Formula: C17H26N8O5HClSolubility: Soluble in water. Clear and faint yellow solution at 5 mg/ml in water. Insoluble in organic solvents.Storage Temperature: +4CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6888