Learn More
AG Scientific Inc Blasticidin S HCl Solution in HEPES Buffer 10 mg/mL, 1X1 mL
Catalog No. NC2734487
Encompass_Preferred
AG Scientific's Blasticidin S HCl solution is a sterile filterate solution. Blasticidin S HCl is an aminoacyl nucleoside antibiotic isolated from Streptomyces griseochromogenes. It functions by inhibiting peptide bond formation at the ribosome in both eukaryotic and prokaryotic cells. It blocks hydrolysis of peptidyl-tRNA due to the large ribosomal subunit's increased affinity for tRNA.Blasticidin S HCL has two deaminase genes: BSD from Aspergillus terreus and bsr from Bacillus cereus, where resistance is shown converting Blasticidin S HCl into a nontoxic deaminohydroxy derivative.Concentration: 10 mg/mL in HEPES buffer
Catalog No. NC2734487 Supplier AG Scientific Inc Supplier No. B1247SOL1ML
AG Scientific's Blasticidin S HCl solution is a sterile filterate solution. Blasticidin S HCl is an aminoacyl nucleoside antibiotic isolated from Streptomyces griseochromogenes. It functions by inhibiting peptide bond formation at the ribosome in both eukaryotic and prokaryotic cells. It blocks hydrolysis of peptidyl-tRNA due to the large ribosomal subunit's increased affinity for tRNA.Blasticidin S HCL has two deaminase genes: BSD from Aspergillus terreus and bsr from Bacillus cereus, where resistance is shown converting Blasticidin S HCl into a nontoxic deaminohydroxy derivative.Concentration: 10 mg/mL in HEPES buffer