Learn More
Research Products International Corp Blasticidin S Hydrochloride 10mg/ml Solution, 200 Milligrams
SDP
Catalog No. 501368107
Encompass_Preferred
Blasticidin S HCl Solution 10 mg/mL is a sterile, DNase/RNase-free solution in HEPES buffer. It is a nucleoside antibiotic derived from Streptomyces griseochromogenes with high purity, commonly used in molecular biology, biotechnology, and gene editing applications such as CRISPR. It functions as a selection agent in cell culture by inhibiting protein synthesis in both prokaryotic and eukaryotic cells, allowing for the isolation of successfully modified cells. This product is ideal for creating stable cell lines and expressing recombinant proteins. Store at -20°C and avoid repeated freeze-thaw cycles.
Catalog No. 50-136-8107 Supplier Research Products International Corp Supplier No. B121500.2
Blasticidin S HCl Solution 10 mg/mL is a sterile, DNase/RNase-free solution in HEPES buffer. It is a nucleoside antibiotic derived from Streptomyces griseochromogenes with high purity, commonly used in molecular biology, biotechnology, and gene editing applications such as CRISPR. It functions as a selection agent in cell culture by inhibiting protein synthesis in both prokaryotic and eukaryotic cells, allowing for the isolation of successfully modified cells. This product is ideal for creating stable cell lines and expressing recombinant proteins. Store at -20°C and avoid repeated freeze-thaw cycles.