Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blasticidin-S, hydrochloride, 99.2%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15047780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15047780 100 mg
ICN15047725 25 mg
ICN15047750 50 mg
MP215047783 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15047780 Supplier MP Biomedicals, Inc Supplier No. 0215047780
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3-9-3513
Molecular Formula C17H27ClN8O5
Molecular Weight (g/mol) 458.90
InChI Key YQXYQOXRCNEATG-UHFFFAOYNA-N
Synonym BLA-S, Cytovirin
SMILES Cl.CN(CCC(N)CC(=O)NC1C=CC(OC1C(O)=O)N1C=CC(N)=NC1=O)C(N)=N

Specifications

Type Blasticidin S Hydrochloride
Physical Form Powder
Color White
Quantity 100 mg
Assay Percent Range 0.992
Formula Weight 458.9
Chemical Name or Material Blasticidin-S hydrochloride, For HPLC analysis
Recommended Storage Store at 4°C.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.