Learn More
Abcam Blasticidin S hydrochloride, Nucleoside antibiotic, 25MG
Catalog No. 502215309
Encompass_Preferred
MW 458.9 Da, Purity >98%. Broad spectrum amino nucleoside antibiotic agent similar to puromycin. Inhibits protein biosynthesis at both prokaryotic and eukaryotic ribosomes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5309 Supplier Abcam Supplier No. AB14145225MG
MW 458.9 Da, Purity >98%. Broad spectrum amino nucleoside antibiotic agent similar to puromycin. Inhibits protein biosynthesis at both prokaryotic and eukaryotic ribosomes.
The product is subject to the following: Abcam Restricted Use Statement