Learn More
Abcam Blasticidin S hydrochloride, Nucleoside antibiotic, 100MG
Catalog No. 502706603
Encompass_Preferred
MW 458.9 Da, Purity >98%. Broad spectrum amino nucleoside antibiotic agent similar to puromycin. Inhibits protein biosynthesis at both prokaryotic and eukaryotic ribosomes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6603 Supplier Abcam Supplier No. AB141452100MG
MW 458.9 Da, Purity >98%. Broad spectrum amino nucleoside antibiotic agent similar to puromycin. Inhibits protein biosynthesis at both prokaryotic and eukaryotic ribosomes.
The product is subject to the following: Abcam Restricted Use Statement