Learn More
Alkali Scientific Blasticidin S Hydrochloride Powder, 1 G
SDP
Supplier:  Alkali Scientific LAS005CP
Encompass_Preferred
Blasticidin S Hydrochloride powder is a potent antibiotic widely used in molecular biology and cell culture applications for selection of genetically modified cells. It inhibits protein synthesis, resulting in rapid death of cells lacking resistance and allowing efficient selection of stable transfectants. This larger pack is suitable for high volume laboratories and extended research use, providing flexibility in preparation of stock solutions at various concentrations. The powder format supports long term storage and consistent performance after reconstitution. It is commonly used in gene expression studies, transfection experiments, and biotechnology workflows requiring strong and reliable antibiotic selection.
Catalog No. 50-006-59911