Learn More
Alkali Scientific Blasticidin S Hydrochloride Powder, 100 Mg
SDP
Catalog No. 5000659902
Encompass_Preferred
Blasticidin S Hydrochloride powder is an effective antibiotic reagent used for selection of genetically modified cells in molecular biology and cell culture workflows. It inhibits protein synthesis, resulting in rapid death of non resistant cells and enabling efficient isolation of stable transfectants. The powder format allows preparation of customized stock solutions and supports long term storage without loss of activity. Once reconstituted, it can be used across a range of biological systems including mammalian and microbial cultures. Researchers commonly use this antibiotic in gene expression studies, transfection protocols, and selection processes requiring strong and reliable antibiotic pressure.
Catalog No. 50-006-59902 Supplier Alkali Scientific Supplier No. LAS003CP
Blasticidin S Hydrochloride powder is an effective antibiotic reagent used for selection of genetically modified cells in molecular biology and cell culture workflows. It inhibits protein synthesis, resulting in rapid death of non resistant cells and enabling efficient isolation of stable transfectants. The powder format allows preparation of customized stock solutions and supports long term storage without loss of activity. Once reconstituted, it can be used across a range of biological systems including mammalian and microbial cultures. Researchers commonly use this antibiotic in gene expression studies, transfection protocols, and selection processes requiring strong and reliable antibiotic pressure.