Learn More
Sigma Aldrich Fine Chemicals Biosciences Blasticidin S Ready Made Solution, 1 mL
Catalog No. NC2002643
Encompass
Blasticidin S belongs to the aminoacylnucleoside class of antibiotics. It inhibits protein synthesis in bacteria and eukaryotes. It also has fungicidal properties and prevents rice blast disease. Blasticidin S inhibits hydrolysis of peptidyl-tRNA induced by release factors. It enhances the binding of tRNA to the large subunit of ribosomes and to an extent inhibits peptide bond formation.
Catalog No. NC2002643 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SBR000221ML
Blasticidin S belongs to the aminoacylnucleoside class of antibiotics. It inhibits protein synthesis in bacteria and eukaryotes. It also has fungicidal properties and prevents rice blast disease. Blasticidin S inhibits hydrolysis of peptidyl-tRNA induced by release factors. It enhances the binding of tRNA to the large subunit of ribosomes and to an extent inhibits peptide bond formation.