Learn More
Cytoskeleton, Inc. BlastR Rapid Lysate Prep Kit (50 filters)
Catalog No. NC3443988
Encompass
The BlastR™ filters allows for fast and reliable preparation of genomic DNA (gDNA)-free cell lysate for western blot or immunoprecipitation when used in combination with a denaturing buffer, such as guanidine, urea or high SDS based extraction buffers that usually produce copious viscosity. gDNA contamination is a significant problem with denaturing buffers that can interfere with downstream applications like immunoprecipitation and migration of proteins in SDS-PAGE. Unlike sonication or insulin needle gDNA shearing methods, the BlastR™ filter effectively removes gDNA contamination while having no effect on the integrity of the proteins in the lysate.
Catalog No. NC3443988 Supplier Cytoskeleton, Inc. Supplier No. BLR01
The BlastR™ filters allows for fast and reliable preparation of genomic DNA (gDNA)-free cell lysate for western blot or immunoprecipitation when used in combination with a denaturing buffer, such as guanidine, urea or high SDS based extraction buffers that usually produce copious viscosity. gDNA contamination is a significant problem with denaturing buffers that can interfere with downstream applications like immunoprecipitation and migration of proteins in SDS-PAGE. Unlike sonication or insulin needle gDNA shearing methods, the BlastR™ filter effectively removes gDNA contamination while having no effect on the integrity of the proteins in the lysate.