Learn More
Bioworld Blaydes Basal Medium, 50 L
Catalog No. 50254840
Encompass_Preferred
Blaydes Basal MediumContains Macronutrients, Micronutrients, Sucrose, and Thiamine as described by Blaydes (1966)Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 31.86Soluble in water
Catalog No. 50-254-840 Supplier Bioworld Supplier No. 306301413
Blaydes Basal MediumContains Macronutrients, Micronutrients, Sucrose, and Thiamine as described by Blaydes (1966)Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 31.86Soluble in water