Learn More
MSC Betco Machine Detergent, Solid, Fresh, 8 lb, 4 Case
Catalog No. 502838617
Encompass
This highly concentrated, 100% active solid detergent is for high and low-temperature dish machines. It includes soil suspension agents and water conditioners to prevent lime deposits, ensuring free and complete rinsing. Its compact, proprietary jar allows for easy handling, quick change-outs, and reduces storage needs. It's economical with 100% active ingredients, ensuring no waste.
  • Concentrated solid machine detergent
  • Compact design with molded handle for easy use and storage
  • Prevents misuse; only for chemical management systems
  • No waste; all product evacuated from capsule
  • Economical (100% active ingredients)
  • Combats challenging water conditions and promotes complete rinsing
  • Prevents lime deposits on dish machine interiors
  • Made in America
Catalog No. 50-283-8617 Supplier MSC Supplier No. 72530223
This highly concentrated, 100% active solid detergent is for high and low-temperature dish machines. It includes soil suspension agents and water conditioners to prevent lime deposits, ensuring free and complete rinsing. Its compact, proprietary jar allows for easy handling, quick change-outs, and reduces storage needs. It's economical with 100% active ingredients, ensuring no waste.
  • Concentrated solid machine detergent
  • Compact design with molded handle for easy use and storage
  • Prevents misuse; only for chemical management systems
  • No waste; all product evacuated from capsule
  • Economical (100% active ingredients)
  • Combats challenging water conditions and promotes complete rinsing
  • Prevents lime deposits on dish machine interiors
  • Made in America