Learn More
Gold Biotechnology Inc Blazin Blue Protein Gel Stain 1 L
SDP
Catalog No. NC2943378
Encompass_Preferred
Blazin' Blue Protein Gel Stain is a ready-to-use protein gel staining solution that is a safer and more effective replacement for traditional Coomassie stains. Blazin' Blue offers a faster alternative to protein staining (5-60 minutes) in a single step without the need for fixation or washing. Protein bands can be detected through visible blue staining or by near-infrared fluorescence. And since Blazin' Blue is aqueous based, there are no hazardous chemicals like methanol or acetic acid to worry about. Blazin' Blue Protein Gel Stain is certified under CCR Title 22 as non-toxic to the environment for drain disposal after a simple pH neutralization step.
Catalog No. NC2943378 Supplier Gold Biotechnology Inc Supplier No. P8101
Blazin' Blue Protein Gel Stain is a ready-to-use protein gel staining solution that is a safer and more effective replacement for traditional Coomassie stains. Blazin' Blue offers a faster alternative to protein staining (5-60 minutes) in a single step without the need for fixation or washing. Protein bands can be detected through visible blue staining or by near-infrared fluorescence. And since Blazin' Blue is aqueous based, there are no hazardous chemicals like methanol or acetic acid to worry about. Blazin' Blue Protein Gel Stain is certified under CCR Title 22 as non-toxic to the environment for drain disposal after a simple pH neutralization step.