Learn More
ABclonal Technology BLBP/FABP7 Rabbit mAb
SDP
Catalog No. 5000005349
Encompass_Preferred
The gene encodes a small highly conserved cytoplasmic protein that bind long-chain fatty acids and other hydrophobic ligands The encoded protein is important in the establishment of the radial glial fiber in the developing brain Alternative splicing and promoter usage results in multiple transcript variants encoding different isoforms Pseudogenes of this gene are found on multiple chromosomes
Catalog No. 50-000-05349 Supplier ABclonal Technology Supplier No. A2328620UL
The gene encodes a small highly conserved cytoplasmic protein that bind long-chain fatty acids and other hydrophobic ligands The encoded protein is important in the establishment of the radial glial fiber in the developing brain Alternative splicing and promoter usage results in multiple transcript variants encoding different isoforms Pseudogenes of this gene are found on multiple chromosomes