Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for Titer Plate, direct contact, 1 Well, 2064Q
SDP
Catalog No. 5032282
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 1 Well
Model: 2064Q
Catalog No. 50-322-82 Supplier Electron Microscopy Sciences Supplier No. 6313301
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Wells: 1 Well
Model: 2064Q