Learn More
Andwin Scientific BLEACH 5.25% SOL
SDP
Catalog No. NC9891719
Encompass_Preferred
The bleach is a yellow liquid chemical that has a chlorine-like odor. It is classified as a base with its pH of 11.5-12.5. This chemical is composed of 94.75% water and 5.25% sodium hypochlorite. It is packaged in a poly bottle. 4X1Gal
This item is not returnable. View return policy
Catalog No. NC9891719 Supplier Andwin Scientific Supplier No. VT150D
The bleach is a yellow liquid chemical that has a chlorine-like odor. It is classified as a base with its pH of 11.5-12.5. This chemical is composed of 94.75% water and 5.25% sodium hypochlorite. It is packaged in a poly bottle. 4X1Gal