Learn More
MSC Clorox Healthcare Bleach Germicidal Wipes, 6 x 5 in, 150 Count, Unscented
Supplier: MSC 65352817
Designed for healthcare professionals, these premoistened wipes effectively disinfect surfaces. They kill C. difficile and C. auris in 3 minutes, and over 60 other microorganisms, including outbreak-causing viruses, emerging viral pathogens, MRSA, and VRE, in 1 minute or less. Their optimized formula provides greater surface coverage with fast contact times and minimal residue.
- Kills C. difficile and C. auris in 3 minutes.
- Kills over 60 microorganisms, including outbreak-causing viruses, emerging viral pathogens, MRSA, and VRE in 3 minutes or less.
- Disinfects more surface area per wipe with fast contact times and greater wetness surface coverage.
- Optimized formula leaves less residue behind.
- Recommended in equipment cleaning and care guides.
- Trusted by over 2,400 US hospitals as part of their infection control protocols.
- EPA Reg. No. 67619-12.