Learn More
MSC Clorox Healthcare Bleach Germicidal Wipes, 1-Ply, 6.75" x 9", Unscented
Catalog No. 502835897
Encompass
Clorox Healthcare Bleach Germicidal Wipes are pre-saturated, 1-ply flat sheets designed for surface disinfecting. These unscented wipes are suitable for various environments including hospitality, restaurant, education, healthcare, high traffic locations, gyms, and office buildings. They come in a resealable pack for convenient use.
  • Disinfecting wipe type
  • Flat sheet form
  • Pre-saturated wipe style
  • Resealable pack packaging
  • White color
  • Unscented
  • Dispensing container included
  • Surface disinfecting application
  • Sheet length: 9 in
  • Sheet width: 6.75 in
Catalog No. 50-283-5897 Supplier MSC Supplier No. 82784190
Clorox Healthcare Bleach Germicidal Wipes are pre-saturated, 1-ply flat sheets designed for surface disinfecting. These unscented wipes are suitable for various environments including hospitality, restaurant, education, healthcare, high traffic locations, gyms, and office buildings. They come in a resealable pack for convenient use.
  • Disinfecting wipe type
  • Flat sheet form
  • Pre-saturated wipe style
  • Resealable pack packaging
  • White color
  • Unscented
  • Dispensing container included
  • Surface disinfecting application
  • Sheet length: 9 in
  • Sheet width: 6.75 in