Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Bleach-Rite Disinfectant Spray
SDP

Catalog No. SBRSPRAYGAL
Change view
Click to view available options
Quantity:
16 oz.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBRSPRAYGAL 4 x 1 gal.
SBRSPRAY16 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. SBRSPRAYGAL Supplier StatLab Supplier No. BRSPRAY/GAL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use pre-mixed bleach concentration.

  • Contains pre-mixed, ready-to-use bleach at a 1:10 concentration at expiration
  • Formulated concentration is recommended by the NCCLS, OSHA, CDC for disinfection against bacteria, viruses, spores
  • Eliminates the need to mix concentrated bleach with water daily
  • EPA approved to kill Coronavirus in 1 minute
  • Gallon refill, 4 per case

Specifications

Product Type Disinfectant
Color-coded No
Form Liquid
pH 12
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Filtered Non-filtered
Flammable No
Quantity 4 x 1 gal.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.