Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLEACH-RITE™ Disinfecting Spray

Catalog No. 17800700
Change view
Click to view available options
Volume (English):
32 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
17-800-700 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-800-700 Supplier Current Tech Inc Supplier No. BRSPRAY32ALT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BLEACH-RITE™ Disinfecting Spray contains pre-mixed, ready-to-use bleach at a 1:10 concentration (0.525% bleach) at expiration, which is the concentration recommended by the NCCLS, OSHA, CDC for disinfection against bacteria, viruses, spores. BLEACH-RITE™ eliminates the need to mix concentrated bleach with water daily.

  • Use to clean/disinfect hard non-porous surfaces and to preclean/decontaminate; not to be used as a sterilant or on any surface or instrument to be introduced directly into the human body.
  • Clinical and Reference labs use BLEACH-RITE to clean/disinfect counters, sinks, and on numerous instruments and analyzers, centrifuges, cryostats (in accordance with their manufacturers' specifications).
  • Research, Biotech and Pharma labs use BLEACH-RITE for disinfection of instruments used in molecular analysis, gene sequencing, and to clean / disinfect centrifuges, cryostats, freezers, counters, sinks.
  • General hospital and physician office areas for use include ER, OR, ICU, radiology, nursing stations, patient rooms, clinics, dialysis and outpatient. Often used to disinfect counters, sinks, blood glucose monitors, outer sides of equipment, IV poles, bed railings, probes, blood pressure cuffs, mammography equipment, carts, wheelchairs, gurneys, EMS backboards, point of care instruments.
  • Veterinary uses include disinfection of animal equipment, cages, treatment areas, counters, sinks. Tests at an independent lab showed that BLEACH-RITE eliminates Canine Parvovirus and Feline Panleukopenia in two minutes, and the EPA approved the claims against these animal-borne viruses.
Note: Case of 6 bottles comes with 3 sprayer heads.

Specifications

Concentration 10:1 / 0.525% bleach
Container Type Spray Bottle
Description Hospital-grade one-step cleaner / Disinfectant
Includes Case of 6 bottles; includes 3 sprayer heads.
Quantity 6/Cs.
Volume (English) 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.