Learn More
Andwin Scientific BLEACH-RITE SPRAY 1 GAL 4/CS
SDP
Catalog No. NC9582575
Encompass_Preferred
The Bleach-Rite disinfecting spray is a ready-to-use cleaning agent that can kill bacteria in a few moments after contact. It is safe to use on several surfaces including plastic, glass, glazed ceramic tile, linoleum, laminated plastic counters, baked enamel, Plexiglas, and glazed porcelain. This product is packaged in a 3.8 L or 1 gal bottle.
This item is not returnable. View return policy
Catalog No. NC9582575 Supplier Andwin Scientific Supplier No. 66025688
The Bleach-Rite disinfecting spray is a ready-to-use cleaning agent that can kill bacteria in a few moments after contact. It is safe to use on several surfaces including plastic, glass, glazed ceramic tile, linoleum, laminated plastic counters, baked enamel, Plexiglas, and glazed porcelain. This product is packaged in a 3.8 L or 1 gal bottle.