Learn More
Cayman Chemical (+)-Blebbistatin, Purity: ≥98%, CAS: 1177356-70-5, 10 mg
SDP
Catalog No. 502955814
Encompass_Preferred
An inactive enantiomer of (–)-blebbistatin; (R)-nitro-blebbistatin is a more stable form of (+)-blebbistatin
Catalog No. 50-295-5814 Supplier Cayman Chemical Supplier No. 1316510 MG
An inactive enantiomer of (–)-blebbistatin; (R)-nitro-blebbistatin is a more stable form of (+)-blebbistatin