Learn More
Medchemexpress LLC HY-13441 10mg Medchemexpress, (-)-Blebbistatin CAS:856925-71-8 Purity:>98%
SDP
Catalog No. 502028995
Encompass_Preferred
Medchemexpress, HY-13441 10mg (-)-Blebbistatin CAS:856925-71-8 (-)-Blebbistatin is a selective inhibitor of the ATPase activity of non-muscle myosin II[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8995 Supplier Medchemexpress LLC Supplier No. HY1344110MG
Medchemexpress, HY-13441 10mg (-)-Blebbistatin CAS:856925-71-8 (-)-Blebbistatin is a selective inhibitor of the ATPase activity of non-muscle myosin II[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.