Learn More
Cayman Chemical BLEBBISTATIN 10MG
SDP
Catalog No. 502953198
Encompass_Preferred
A cell-permeable inhibitor of non-muscle myosin II ATPases; inhibits Mg-ATPase activity and in vitro motility of non-muscle myosin IIA and IIB for several species (IC50s = 0.5-5 µM); poorly inhibits smooth muscle myosin (IC50 = 80 µM); (S)-nitro-blebbistatin is a more stable form of (-)-Blebbistatin
Catalog No. 50-295-3198 Supplier Cayman Chemical Supplier No. 1301310 MG
A cell-permeable inhibitor of non-muscle myosin II ATPases; inhibits Mg-ATPase activity and in vitro motility of non-muscle myosin IIA and IIB for several species (IC50s = 0.5-5 µM); poorly inhibits smooth muscle myosin (IC50 = 80 µM); (S)-nitro-blebbistatin is a more stable form of (-)-Blebbistatin