Learn More
Cayman Chemical (±)-Blebbistatin, 674289-55-5, 25mg
SDP
Supplier:  Cayman Chemical 1318625
Encompass_Preferred
Molecular Formula C18H16N2O2, Purity ≥98%, Formula Weight 292.3, Synonyms: 1,2,3,3a-tetrahydro-3a-hydroxy-6-methyl-1-phenyl-4H-pyrrolo[2,3-b]quinolin-4-one; A racemic mixture of blebbistatin isomers.
Catalog No. NC0873310