Learn More
Abcam (±)-Blebbistatin, inhibitor of myosin II, 5MG
Supplier:  Abcam AB1204255MG
Encompass_Preferred
MW 292.3 Da, Purity >99%. Cell-permeable, reversible inhibitor of non-muscle myosin II. Shows high affinity and selectivity for vertebrate and invertebrate Myosin II (IC50 values 0.5-5 μM). Inhibits actin-activated MgATPase activity and *in vitro* motility of class II myosins. Also inhibits contraction of the cytokinetic ring. Blocks cell blebbing and disrupts directed cell migration and cytokinesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1327356