Learn More
Apexbio Technology LLC (-)-Blebbistatin 856925-71-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13875.1
Encompass_Preferred
(-)-Blebbistatin is a cell-permeable small molecule inhibitor targeting non-muscle myosin II (NM II) an actin-dependent motor protein involved in cell adhesion migration and differentiation It selectively binds the myosin-ADP-phosphate complex slowing phosphate release and thereby suppressing Mg-ATPase and contractile activities mediated by actomyosin interactions It exhibits inhibitory potency in various model systems (IC50 0 5 5 0 M) with minimal effects on myosin isoforms I V X and reduced activity toward smooth muscle myosin II (IC50 80 M) Its reversible modulation of NM II renders it useful in research on cytoskeletal dynamics cell mechanics and related pathophysiological conditions
Catalog No. 50-000-63471