Learn More
Selleck Chemical LLC (-)-Blebbistatin 25mg 856925-71-8
SDP
Supplier:  Selleck Chemical LLC S709925MG
Encompass_Preferred
(-)-Blebbistatin is a cell-permeable inhibitor for non muscle myosin II ATPase with IC50 of ~2 ?M in cell-free assays, does not inhibit myosin light chain kinase, inhibits contraction of the cleavage furrow without disrupting mitosis or contractile ring assembly. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4404