Learn More
TARGETMOL CHEMICALS INC --BLEBBISTATIN 25MG
SDP
Catalog No. 502579623
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Blebbistatin ((S)-(-)-Blebbistatin) is an S enantiomer of blebbistatin. It is a potent and selective myosin II inhibitor with IC50 ranging from 0.5 to 5 uM. purity: 99%
Catalog No. 50-257-9623 Supplier TARGETMOL CHEMICALS INC Supplier No. T603825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Blebbistatin ((S)-(-)-Blebbistatin) is an S enantiomer of blebbistatin. It is a potent and selective myosin II inhibitor with IC50 ranging from 0.5 to 5 uM. purity: 99%