Learn More
Enzo Life Sciences Blebbistatin(+/-), (5mg), CAS Number: 674289-55-5
SDP
Catalog No. NC9051761
Encompass_Preferred
Inhibitor of non-muscle myosin II. Blebbistatin (so named because of its ability to block cell blebbing) is a selective and high-affinity (IC50 approx. 4 µM) inhibitor of non-muscle myosin II. During cell division blebbistatin inhibits contraction of the cleavage furrow without disrupting mitosis. It inhibits pancreatic adenocarcinoma cellular invasiveness and PDGF-stimulated fibroblast-collagen matrix contraction. Formula: C18H16N2O2. MW: 292.3. CAS Number: 674289-55-5. Purity: ≥97%. Appearance: Bright yellow solid. Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C
Catalog No. NC9051761 Supplier Enzo Life Sciences Supplier No. BMLEI3150005
Inhibitor of non-muscle myosin II. Blebbistatin (so named because of its ability to block cell blebbing) is a selective and high-affinity (IC50 approx. 4 µM) inhibitor of non-muscle myosin II. During cell division blebbistatin inhibits contraction of the cleavage furrow without disrupting mitosis. It inhibits pancreatic adenocarcinoma cellular invasiveness and PDGF-stimulated fibroblast-collagen matrix contraction. Formula: C18H16N2O2. MW: 292.3. CAS Number: 674289-55-5. Purity: ≥97%. Appearance: Bright yellow solid. Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C