Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America™ Bleed Control Trauma Management Station
SDP

Catalog No. 19777002
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-777-002 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-777-002 Supplier Ready America™ Supplier No. 10360
  • $617.00
    $459.65 / Each of 1

    Save $157.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready America™ Bleed Control Trauma Management Station is wall-mounted break glass cabinet that contains a range of critical medical supplies for bleed control response.

  • Ideal for grade schools, universities, offices, warehouses, or places of worship
  • Includes the following:
    • 1 Pair of Safety Glasses
    • 2 Pair of Nitrile Gloves
    • 2 N-95 Mask
    • 2 Survival Blanket
    • 1 CPR Mouth Shield
    • 1 Kerlic Gauze Roll
    • 1 Emergency Pressure Dressing
    • 2 Multi Trauma Dressing
    • 1 Blood Stopper Trauma Dressing
    • 4 Surgi Pad Combine Dressing
    • 4 4 in. x 4.1 yds Roll Gauze Roll
    • 4 Rapid Response Wound Seal
    • 1 EMT Paramedic Scissors
    • 2 Surgical Adhesive Tape
    • 2 Triangular Bandage
    • 1 Duct Tape 10 Yards
    • 1 Doctor’s Pen Light
    • 1 Hydrogen Peroxide
    • 2 Bio Hazard Bags
    • 1 Emergency High-Strength Pressure Bandage, 6 in.
    • 1 C-A-T Tourniquet
    • 1 QuikClot™ Bleeding Control DressingTM
    • 1 Sharpie
    • 1 Saline Solution (Sodium Chloride), 500ml Bottle
    • 1 Ammonia Inhalants, Box of 10

Specifications

Product Type First Aid Kit
For Use With (Application) Bleed Control Management
Height (English) 9 in.
Length (English) 22 in.
Length (Metric) 55.88 cm
Color Red
Quantity 1 Ea.
Width (English) 10 in.
Width (Metric) 25.4 cm
Height (Metric) 22.86 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.