Learn More
Grainger Blender 1 gal 3-3/4 HP
Catalog No. 502764335
Encompass_Preferred
The Waring Commercial Blender is a heavy-duty commercial blender with a 128 oz polycarbonate container. It has a 3 peak output horsepower motor and stainless steel blades. This blender has variable speed control with high, low and pulse settings. It has a vented lid with a removable measuring cap. The base is made of die-cast metal and has non-slip rubber feet. This blender is NSF certified, ETL listed and made in the USA. It comes with a 3 year warranty on motor base and 1 year warranty on jar and parts.
Catalog No. 50-276-4335 Supplier Grainger Supplier No. 16W645
The Waring Commercial Blender is a heavy-duty commercial blender with a 128 oz polycarbonate container. It has a 3 peak output horsepower motor and stainless steel blades. This blender has variable speed control with high, low and pulse settings. It has a vented lid with a removable measuring cap. The base is made of die-cast metal and has non-slip rubber feet. This blender is NSF certified, ETL listed and made in the USA. It comes with a 3 year warranty on motor base and 1 year warranty on jar and parts.