Learn More
Hach Company Blender-Two Speed-1.2L
Supplier:  Hach Company 2616100
Encompass
Blender-Two Speed-1 2-L-120 Vac-50/60 Hz This blender is tough enough to handle homogenizing wastewater sludge soil or heavy weight food samples for superior analytical results Both models have thick walled glass containment jars and durable rust free stainless steel blades Two-piece vinyl and styrene lid
Catalog No. 50-002-57143