Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bleomycin A5 Hydrochloride, G-Biosciences
SCI_ED

Catalog No. S139842
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139842 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139842 Supplier G-Biosciences Supplier No. RC899
Add to Cart
Add to Cart

Bleomycin A5 hydrochloride is a compound part of the glycopeptide antibiotic family.

Bleomycin A5 stimulates downstream apoptosis by increasing cysteine-aspartic acid protease-3 (caspase-3) activity, tumor protein 53 (p53) production and transforming growth factor beta 1 (TGF-beta 1) formation. These proteins induce apoptosis directly or indirectly by halting cell growth.

Chemical Identifiers

Molecular Weight (g/mol) 1477.02 g/mol

Specifications

Quantity 10 mg
Percent Purity 95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.