Learn More
Medchemexpress LLC HY-17565A 10mg Medchemexpress, Bleomycin (hydrochloride) CAS:67763-87-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17565A10MG
Encompass_Preferred
Medchemexpress, HY-17565A 10mg Bleomycin (hydrochloride) CAS:67763-87-5 Bleomycin hydrochloride is a DNA synthesis inhibitor. Bleomycin hydrochloride is a DNA damaging agent. Bleomycin hydrochloride is an antitumor antibiotic. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0791
 Restricted Item