Learn More
Medchemexpress LLC Bleomycin (hydrochloride) | 67763-87-5 | 99.5% | 5 MG
SDP
Catalog No. 5000387150
Encompass_Preferred
Bleomycin hydrochloride is a DNA synthesis inhibitor, a DNA damaging agent, and an antitumor antibiotic. It is primarily used for research purposes, including the creation of animal models of pulmonary fibrosis.
  • Inhibits DNA synthesis.
  • Acts as a DNA damaging agent.
  • Functions as an antitumor antibiotic.
  • Useful for creating animal models of pulmonary fibrosis.
Catalog No. 50-003-87150 Supplier Medchemexpress LLC Supplier No. HY17565A5MG
Bleomycin hydrochloride is a DNA synthesis inhibitor, a DNA damaging agent, and an antitumor antibiotic. It is primarily used for research purposes, including the creation of animal models of pulmonary fibrosis.
  • Inhibits DNA synthesis.
  • Acts as a DNA damaging agent.
  • Functions as an antitumor antibiotic.
  • Useful for creating animal models of pulmonary fibrosis.