Learn More
Cusabio Technology LLC bleomycin hydrolase
Catalog No. 5000051660
Encompass_Preferred
The Human Bleomycin Hydrolase (BLMH) ELISA Kit is designed to quantitatively measure BLMH protein levels in diverse biological samples including serum plasma tissue homogenates and cell lysates Utilizing a sandwich ELISA methodology this assay demonstrates a detection range of 28 pg/mL to 1800 pg/mL with a sensitivity of 7 pg/mL enabling precise measurement of low-abundance targets The procedure requires 50-100 L of sample per test and can be completed within 1-5 hours with absorbance read at 450 nm for result determination Its robust performance makes it suitable for research applications requiring reliable quantification of BLMH expression in both normal and pathological conditions
Catalog No. 50-000-51660 Supplier Cusabio Technology LLC Supplier No. CSBEL002716HU
The Human Bleomycin Hydrolase (BLMH) ELISA Kit is designed to quantitatively measure BLMH protein levels in diverse biological samples including serum plasma tissue homogenates and cell lysates Utilizing a sandwich ELISA methodology this assay demonstrates a detection range of 28 pg/mL to 1800 pg/mL with a sensitivity of 7 pg/mL enabling precise measurement of low-abundance targets The procedure requires 50-100 L of sample per test and can be completed within 1-5 hours with absorbance read at 450 nm for result determination Its robust performance makes it suitable for research applications requiring reliable quantification of BLMH expression in both normal and pathological conditions