Learn More
Sigma Aldrich Fine Chemicals Biosciences Bleomycin ready made solut
Catalog No. 502905520
Encompass
Bleomycin (BLM) is a glycopeptide antibiotic consist of 2 main isomers BLM A2 and B2 obtained from Streptomyces verticillus. The main clinical application of Bleomycin is cancer treatment. Bleomycin causes oxidative degradation of RNA affecting G2 and M cell cycle phases. The anti-cancer mechanism of Bleomycin is mediated through iron chelation and generation of oxygen free radicals leading to DNA cleavage and ultimately to cell death. Bleomycin hydrolase the enzyme responsible for Bleomycin metabolism is normally present at low levels in the lungs. Therefore Bleomycin treatment may result in its accumulation in the lungs and can potentially cause lung injury due to lung cell death. The lung injury occurs through overproduction of reactive oxygen species causing pulmonary toxicity leading to fibrosis. The lung fibrosis by Bleomycin can be utilized for screening of antifibrotic agents.Bleomycin has several important applicationsBleomycin has been used to induce chronic lung d
Catalog No. 50-290-5520 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B72161ML
Bleomycin (BLM) is a glycopeptide antibiotic consist of 2 main isomers BLM A2 and B2 obtained from Streptomyces verticillus. The main clinical application of Bleomycin is cancer treatment. Bleomycin causes oxidative degradation of RNA affecting G2 and M cell cycle phases. The anti-cancer mechanism of Bleomycin is mediated through iron chelation and generation of oxygen free radicals leading to DNA cleavage and ultimately to cell death. Bleomycin hydrolase the enzyme responsible for Bleomycin metabolism is normally present at low levels in the lungs. Therefore Bleomycin treatment may result in its accumulation in the lungs and can potentially cause lung injury due to lung cell death. The lung injury occurs through overproduction of reactive oxygen species causing pulmonary toxicity leading to fibrosis. The lung fibrosis by Bleomycin can be utilized for screening of antifibrotic agents.Bleomycin has several important applicationsBleomycin has been used to induce chronic lung d